Please login to your online account to display your discounted pricing

Dowex™ 1X8-100, ion-exchange resin, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  202991000

 View more versions of this product

Catalog No. AC202991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder and/or Chunks
Quantity 100g
Color White
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Chemical Name or Material Dowex™ 1X8
Particle Size 15 % max. (on 35 mesh, cumulative) 15 % max. (through 80 mesh)
Packaging Plastic bottle
Water Volume Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 43 to 48%
MDL Number MFCD00132718
CAS 12627-85-9

Specifications

Chemical Name or Material Dowex™ 1X8
CAS 12627-85-9
MDL Number MFCD00132718