Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 100-200 (Cl)

$53.73 - $660.97

Chemical Identifiers

Chemical Name or Material Dowex™ 1X8 100-200 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL142570B

  SDS
alfa aesar™
L142570B
1000g Each for $660.97

AAL1425718

  SDS
alfa aesar™
L1425718
50g Each for $53.73

AAL1425730

  SDS
alfa aesar™
L1425730
250g Each for $202.46
Description & Specifications

Specifications

CAS 12627-85-9
MDL Number MFCD00132718
Quantity 1000g
Odor Amine-like
Chemical Name or Material Dowex™ 1X8 100-200 (Cl)

Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.