Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 100-200 (Cl)


Quantity 50g

Manufacturer: alfa aesar™  L1425718

 View more versions of this product

Catalog No. AAL1425718

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Dowex™ 1X8 100-200 (Cl)
Odor Amine-like
MDL Number MFCD00132718
Quantity 50g
CAS 12627-85-9

Specifications

Chemical Name or Material Dowex™ 1X8 100-200 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718

Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.