missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AmberChrom 1x2 50-100 (Cl), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1213030
Description
It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
Dowex™ 1X2 50-100 (Cl) | |
Amine-like | |
MFCD00132715 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride | |
16212807 |
9085-42-1 | |
C29H34ClN | |
250 g | |
Insoluble in water. | |
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 | |
432.05 |
Chemical Identifiers
9085-42-1 | |
432.05 | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
16212807 |
C29H34ClN | |
MFCD00132715 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only