missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 1X2-400, ion-exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202971000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinSpecifications
Beads | |
9085-42-1 | |
Plastic bottle | |
C29H34ClN | |
Cl form: 70 to 80% | |
10, 2182 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride | |
16212807 |
Dowex™ 1X2 | |
Particle size analysis: on 70 Mesh (cumulative): <15%, through 270 Mesh: <15% | |
wet volume capacity, Cl form (meq/mL):> 0.6 | |
MFCD00132715 | |
01,511; 04,267; 06,302; 07,182; 08,263 | |
100 g | |
Solubility in water: insoluble. | |
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 | |
432.05 |
Chemical Identifiers
9085-42-1 | |
432.05 | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
16212807 |
C29H34ClN | |
MFCD00132715 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 |
RUO – Research Use Only