Please login to your online account to display your discounted pricing

Dowex™ 1X2-200, ion-exchange resin, ACROS Organics™


Quantity 100g
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  202961000

 View more versions of this product

Catalog No. AC202961000

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Merck Index 10, 2182
Physical Form Beads
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Particle Size Particle size analysis: on 50 mesh (cumulative): < 15%, through 140 mesh: < 15%
Packaging Plastic Bottle
Water Volume Capacity wet volume capacity, Cl form (meq/mL):> 0.6
Water Retention Capacity Cl form: 70 to 80%
MDL Number MFCD00132715
CAS 9085-42-1
Chemical Name or Material Dowex™ 1X2

Specifications

Chemical Name or Material Dowex™ 1X2
CAS 9085-42-1
MDL Number MFCD00132715