Dow Corning™ Silastic™ Laboratory Tubing
Flexible, translucent silicone elastomer tubing
Manufacturer:  Dow Corning Corp 2415666
 View more versions of this product
Catalog No. 11-189-13B
Specifications Description & Specifications
Specifications
2.38mm (0.094 in.)
-65.2° to 480°F
Fluid processing, separation and filtration such as drug manufacturing, oxygen delivery in production processes and chemical, pharmaceutical and environmental analyses
0.094 in.
-54° to 249°C
Translucent
0.31 in.
7.8mm
0.5 in.
Peristaltic pump
12.7mm
50 ft.
15.3m
ISO 9001, FDA, USP Class VI
Yes
Silicone
Accessories