Please login to your online account to display your discounted pricing
American Educational Products Double Convex Unmounted Lens
$7.10 - $8.65
Specifications
Diameter (English) 1.496 in.
Diameter (Metric) 3.8cm
Product Type Double Convex Unmounted Lens
View More Specs
Products
Catalog Number Mfr. No. Length (English) Focal Length (Metric) Focal Price Quantity    
S95358
american educational products
7-909-4
7.87 in. 20cm Each for $7.10
Dozen of 12 Each for $85.05
S95357
american educational products
7-909-3
5.9 in. 15cm Each for $8.65
Dozen of 12 Each for $103.80
Description & Specifications
Specifications
Diameter (English) 1.496 in.
Diameter (Metric) 3.8cm
Product Type Double Convex Unmounted Lens