missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dotriacontane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC117690050
Specifications
Dotriacontane | |
Authentic | |
467°C | |
67°C to 72°C | |
97% | |
97% | |
CH3(CH2)30CH3 | |
4-01-00-00595 beilstein handbook reference, acmc-1akq8, bicetyl, dicetyl, dotriacontane, dotriacontane, n, lacceran, n-dotriacontane, unii-7ksv90rn23, wln: 32h | |
QHMGJGNTMQDRQA-UHFFFAOYSA-N | |
dotriacontane | |
11008 | |
450.86 | |
97% |
247°C | |
Glass bottle | |
Gray to White | |
5g | |
544-85-4 | |
C32H66 | |
MFCD00009411 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in ethyl ether,very soluble in benzene | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
450.86 | |
CHEBI:36020 | |
Shiny Flakes |
Chemical Identifiers
544-85-4 | |
450.86 | |
QHMGJGNTMQDRQA-UHFFFAOYSA-N | |
11008 | |
dotriacontane |
C32H66 | |
MFCD00009411 | |
4-01-00-00595 beilstein handbook reference, acmc-1akq8, bicetyl, dicetyl, dotriacontane, dotriacontane, n, lacceran, n-dotriacontane, unii-7ksv90rn23, wln: 32h | |
CHEBI:36020 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 208-881-5
RTECSNumber : JT2360000
TSCA : TSCA