missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dotriacontane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 117691000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dotriacontane | |
544-85-4 | |
247°C | |
97% | |
C32H66 | |
MFCD00009411 | |
n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h | |
QHMGJGNTMQDRQA-UHFFFAOYSA-N | |
dotriacontane | |
11008 | |
450.86 |
97% | |
467°C | |
Authentic | |
Plastic bottle | |
CH3(CH2)30CH3 | |
100g | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in ethyl ether,very soluble in benzene | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
450.88 | |
CHEBI:36020 | |
97% |
Safety and Handling
EINECSNumber : 208-881-5
RTECSNumber : JT2360000
TSCA : TSCA
RUO â Research Use Only