Please login to your online account to display your discounted pricing

Dorsomorphin dihydrochloride, Tocris

Potent and selective AMPK inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Dorsomorphin dihydrochloride
Assay Percent Range >98%
Molecular Formula C24H25N5O·2HCl
Formula Weight 472.41
Synonym 6-[4-[2-(1-Piperidinyl)ethoxy]phenyl]-3-(4-pyridinyl)-pyrazolo[1, 5-a]pyrimidine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

309310

 
r & d systems
RDS309310
10mg Each for $239.63

309350

 
r & d systems
RDS309350
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 472.41
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C24H25N5O·2HCl
Chemical Name or Material Dorsomorphin dihydrochloride
Recommended Storage Desiccate at Room Temperature
Synonym 6-[4-[2-(1-Piperidinyl)ethoxy]phenyl]-3-(4-pyridinyl)-pyrazolo[1, 5-a]pyrimidine dihydrochloride
CAS 1219168-18-9
Safety and Handling

Desiccate at Room Temperature