missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Donitriptan hydrochloride

5-HT1B/1D agonist

$192.67 - $812.12

Specifications

Product Type Donitriptan hydrochloride
Inhibitors Donitriptan hydrochloride
Molecular Weight 439.94
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
366510R Tocris™
366510
10mg Each for $192.67
366550R Tocris™
366550
50mg Each for $812.12
Specifications

Description

Brain penetrant 5-HT1B/1D agonist (pKi values are 9.3 and 9.4 for 5-HT1D and 5-HT1B respectively). Inhibits capsaicin-induced external carotid vasodilation and produces selective carotid vasoconstriction in various animal species. Displays antimigraine activity.
  • Chemical Name: 1-[[[3-(2-Aminoethyl)-1H-indol-5-yl]oxy]acetyl]-4-(4-cyanophenyl)-piperazine hydrochloride
  • Formula: C23H25N5O2.HCl
  • CAS Number: 170911-68-9
  • Solubility: Soluble to 50mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

Donitriptan hydrochloride
Donitriptan hydrochloride
439.94
>98