missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Domperidone

Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier

Manufacturer:  Alfa Aesar™ J63681MC

 View more versions of this product

Catalog No. AAJ63681MC



Description

Description

Applications
Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

Domperidone
243°C
57808-66-9
MFCD00069256
domperidon, domperidona, domperidona inn-spanish, domperidone, domperidonum, domperidonum inn-latin, motilium, motillium, motinorm, nauzelin
FGXWKSZFVQUSTL-UHFFFAOYSA-N
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
3151
425.91
White
100mg
C22H24ClN5O2
14,3418
Slightly soluble in water.
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O
425.917
CHEBI:31515
Crystalline Powder

Chemical Identifiers

57808-66-9
425.917
FGXWKSZFVQUSTL-UHFFFAOYSA-N
3151
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
C22H24ClN5O2
MFCD00069256
domperidon, domperidona, domperidona inn-spanish, domperidone, domperidonum, domperidonum inn-latin, motilium, motillium, motinorm, nauzelin
CHEBI:31515
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H361

EINECSNumber : 260-968-7

RTECSNumber : DE2275900

TSCA : No

Recommended Storage : Ambient temperatures

Documents