Please login to your online account to display your discounted pricing

Domoic acid, Tocris


Potent, selective kainate agonist

Quantity 1mg

Manufacturer: r & d systems  02691

 View more versions of this product

Catalog No. 02691

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1mg
Formula Weight 311.33
Chemical Name or Material Domoic acid
Assay Percent Range >95%
Molecular Formula C15H21NO6
CAS 14277-97-5
Synonym (2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Domoic acid
Assay Percent Range >95%
Molecular Formula C15H21NO6
Formula Weight 311.33
Synonym (2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
Accessories

Desiccate at -20°C

SDS & Safety Documents