Please login to your online account to display your discounted pricing

Domoic acid, Tocris

Potent, selective kainate agonist

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material Domoic acid
Assay Percent Range >95%
Molecular Formula C15H21NO6
Formula Weight 311.33
Synonym (2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

02691

  SDS
r & d systems
02691
1mg Each for $208.25

02695

  SDS
r & d systems
02695
5mg Each for $867.95
Description & Specifications

Specifications

Quantity 1mg
Formula Weight 311.33
Assay Percent Range >95%
Chemical Name or Material Domoic acid
Recommended Storage Desiccate at -20°C
CAS 14277-97-5
Synonym (2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
Molecular Formula C15H21NO6
Safety and Handling

Desiccate at -20°C