Please login to your online account to display your discounted pricing

Thermo Scientific™ Domed PCR Cap Strips


Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.

Manufacturer: thermo scientific™  AB0852

 View more versions of this product

Catalog No. AB-0852

  • / Case of 80 Each
 

Description & Specifications

Specifications

Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
No. per Pack 8
Material Virgin Medical Grade Polypropylene
Color Natural
For Use With 96-well PCR plates as well as standard and low profile PCR Tube Strips
Applications Maximizes contact with heated cycler lid
Quantity 10 packs of 8
Well Count 96
No. per Case 80
Format Strips of 12 Domed Caps

These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.

Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.

Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.