Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Supplier:  Thermo Scientific™ AB0784
 View more versions of this product
Catalog No. AB-0784
Description
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Specifications
Specifications
Clear
8
Strips of 8 Flat Caps
Yes
Polypropylene
250/Cs.
PCR Plate Cap Strip
96-well Plate