Please login to your online account to display your discounted pricing

Dofetilide, Tocris™

KV11.1 (hERG) channel blocker; inhibits rapid delayed rectifier K+ current (IKr)

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material Dofetilide
Assay Percent Range >99%
Molecular Formula C19H27N3O5S2
Formula Weight 441.56
Synonym 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

375710

 
r & d systems
RDS375710
10mg Each for $205.70

375750

 
r & d systems
RDS375750
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 441.56
Quantity 10mg
Assay Percent Range >99%
CAS 115256-11-6
Chemical Name or Material Dofetilide
Synonym 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane
Molecular Formula C19H27N3O5S2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature