Learn More
Dodecyltrimethylammonium bromide, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 409310250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Dodecyltrimethylammonium bromide | |
246.0°C | |
Authentic | |
Glass bottle | |
CH3(CH2)11N(Br)(CH3)3 | |
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
dodecyl(trimethyl)azanium;bromide | |
14249 | |
308.34 | |
Pure |
1119-94-4 | |
White to Yellow | |
98.5% min. (Argentometry) | |
C15H34BrN | |
25 g | |
(10% in water) Clear to slightly hazy | |
CCCCCCCCCCCC[N+](C)(C)C.[Br-] | |
308.34 | |
CHEBI:282662 | |
99% | |
Powder |
Chemical Identifiers
1119-94-4 | |
308.34 | |
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
CHEBI:282662 | |
CCCCCCCCCCCC[N+](C)(C)C.[Br-] |
C15H34BrN | |
XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
14249 | |
dodecyl(trimethyl)azanium;bromide |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
EINECSNumber : 214-290-3
RUO – Research Use Only