Please login to your online account to display your discounted pricing

Dodecyltrimethylammonium bromide, 99%, pure, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC409315000

 View more versions of this product

Catalog No. AC409315000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. Very toxic to aquatic life.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.
Formula Weight 308.34
Assay ≥98.5%
Physical Form Powder
Quantity 500g
Color White to slightly yellow
Purity Grade Pure
Color Index No. 1,2,3
Appearance of Solution (10% aq. soln.) clear to slightly hazy
Health Hazard 1 Warning
Molecular Formula C15H34BrN
Assay Percent Range 99%
CAS 1119-94-4
Chemical Name or Material Dodecyltrimethylammonium bromide
Infrared Spectrum Authentic
Linear Formula CH3(CH2)11N(Br)(CH3)3

Specifications

Chemical Name or Material Dodecyltrimethylammonium bromide
CAS 1119-94-4
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C15H34BrN
Linear Formula CH3(CH2)11N(Br)(CH3)3
Formula Weight 308.34
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.

GHS H Statement
May cause respiratory irritation. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. Very toxic to aquatic life.

SDS & Safety Documents