missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dodecylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117661000

 View more versions of this product

Catalog No. AC117661000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dodecylamine
124-22-1
White
247.0°C to 249.0°C
Authentic
Glass bottle
CH3(CH2)11NH2
100 g
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
JRBPAEWTRLWTQC-UHFFFAOYSA-N
dodecan-1-amine
13583
98%
98%
28.0°C to 32.0°C
0.8
>110°C
97.5% min. (Alkyl chain distribution C12) (GC)
C12H27N
MFCD00008154
04, 200
Solubility in water: insoluble. Other solubilities: soluble in chloroform, isopropylalcohol, ethanol,, ether
CCCCCCCCCCCCN
185.36
185.35
Crystalline Low Melting Solid

Chemical Identifiers

124-22-1
185.36
JRBPAEWTRLWTQC-UHFFFAOYSA-N
13583
CCCCCCCCCCCCN
C12H27N
MFCD00008154
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
dodecan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquat

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 204-690-6

RTECSNumber : JR6475000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only