Learn More
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 226140025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dodecyl sulfate, sodium salt | |
151-21-3,7757-82-6,7647-14-5 | |
206.0°C | |
C12H25NaO4S | |
MFCD00036175 | |
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate | |
>150°C | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
85% | |
Pure |
White to Yellow | |
Powder or Crystals | |
85% | |
CH3(CH2)11OSO3Na | |
2.5 kg | |
Solubility in water: 130g/L (20°C). | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
288.38 | |
Authentic | |
Plastic drum |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium dodecyl sulfate |
C12H25NaO4S | |
MFCD00036175 | |
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Danger
RUO – Research Use Only