missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dodecyl Methacrylate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America M0083500ML

 View more versions of this product

Catalog No. M0083500ML



Specifications

Specifications

Dodecyl Methacrylate (stabilized with MEHQ)
Colorless
142-90-5
MFCD00008972
dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
CCCCCCCCCCCCOC(=O)C(=C)C
254.414
254.41
≥97.0% (GC)
167°C
500mL
C16H30O2
3082
GMSCBRSQMRDRCD-UHFFFAOYSA-N
dodecyl 2-methylprop-2-enoate
8906
Clear Liquid at 20°C

Chemical Identifiers

142-90-5
254.414
GMSCBRSQMRDRCD-UHFFFAOYSA-N
8906
CCCCCCCCCCCCOC(=O)C(=C)C
C16H30O2
MFCD00008972
dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
dodecyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1039

RTECSNumber : OZ4300000

TSCA : Yes

Documents