Please login to your online account to display your discounted pricing

Dodecyl aldehyde, 92%, stabilized, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365225000

 View more versions of this product

Catalog No. AC365225000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥90% (GC)
Physical Form Fine Crystalline Powder
Formula Weight 184.32
Quantity 500mL
Color White to off-white
Density 0.8310g/mL
Beilstein 01, 714
Packaging Glass bottle
Linear Formula CH3(CH2)10CHO
CAS 112-54-9
Refractive Index 1.4330 to 1.4400 (20°C, 589nm)
Stabilizer abt. 0.1% alpha-tocopherol or 250ppm BHT
Chemical Name or Material Dodecyl aldehyde
Assay Percent Range 92%
Molecular Formula C12H24O
MDL Number MFCD00007017
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Synonym Lauraldehyde
Free Acid 2.5% max.
Name Note Stabilized
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dodecyl aldehyde
Name Note Stabilized
CAS 112-54-9
Assay ≥90% (GC)
Assay Percent Range 92%
Molecular Formula C12H24O
Linear Formula CH3(CH2)10CHO
Formula Weight 184.32
MDL Number MFCD00007017
Synonym Lauraldehyde
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

SDS & Safety Documents