Please login to your online account to display your discounted pricing

Dodecyl aldehyde, 92%, stabilized, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dodecyl aldehyde
Name Note Stabilized
CAS 112-54-9
Assay ≥90% (GC)
Assay Percent Range 92%
Molecular Formula C12H24O
Linear Formula CH3(CH2)10CHO
Formula Weight 184.32
MDL Number MFCD00007017
Synonym Lauraldehyde
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365225000

  SDS
thermo fisher scientific
AC365225000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥90% (GC)
Formula Weight 184.32
Physical Form Fine Crystalline Powder
Quantity 500mL
Color White to off-white
Density 0.8310g/mL
Beilstein 01, 714
Packaging Glass bottle
Linear Formula CH3(CH2)10CHO
Stabilizer abt. 0.1% alpha-tocopherol or 250ppm BHT
Refractive Index 1.4330 to 1.4400 (20°C, 589nm)
CAS 112-54-9
Assay Percent Range 92%
Chemical Name or Material Dodecyl aldehyde
Molecular Formula C12H24O
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
MDL Number MFCD00007017
Synonym Lauraldehyde
Free Acid 2.5% max.
Infrared Spectrum Authentic
Name Note Stabilized
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Warning

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.