missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dodecanonitrile, 98%

$592.38 - $592.38

Chemical Identifiers

CAS 2437-25-4
Molecular Formula CH3(CH2)10CN
Formula Weight 181.32
MDL Number MFCD00001946
Beilstein 970348
Synonym Lauronitrile; Undecyl cyanide
Chemical Name or Material Dodecanonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1903830 SDS Alfa Aesar™
A1903830
250g Each for $592.38
AAA1903809 SDS Alfa Aesar™
A1903809
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1903818 SDS Alfa Aesar™
A1903818
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Dodecanonitrile in xylene is used as a solvent and in the presence of sulfur was reduced with molybdenum sulfide catalyst yielded 70-5% dodecylmercaptan.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2437-25-4
181.32
970348
Dodecanonitrile
CH3(CH2)10CN
MFCD00001946
Lauronitrile; Undecyl cyanide

Specifications

250g
CH3(CH2)10CN
MFCD00001946
Lauronitrile; Undecyl cyanide
4°C
Dodecanonitrile
2437-25-4
181.32
970348
272° to 274°C
114°C (237°F)
98%

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : JR2600000

TSCA : TSCA