Please login to your online account to display your discounted pricing

Alfa Aesar™ Dodecanoic hydrazide, 97%

$694.46 - $694.46

Chemical Identifiers

Chemical Name or Material Dodecanoic hydrazide
CAS 5399-22-4
Molecular Formula CH3(CH2)·CONHNH2
Formula Weight 214.35g/mol
MDL Number MFCD00025132
Synonym Lauric acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2515114

 
alfa aesar™
B25151-14
25g Each for $694.46
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Dodecanoic hydrazide
CAS 5399-22-4
Assay 97%
Molecular Formula CH3(CH2)·CONHNH2
Formula Weight 214.35g/mol
MDL Number MFCD00025132
Synonym Lauric acid hydrazide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Dodecanoic hydrazide is used as pharmaceutical intermediate. It is used as corrosion inhibitors.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.