missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Dodecane, 99%, pure, ACROS Organics™
Manufacturer: Fisher Scientific Acros 117595000
Specifications
n-Dodecane | |
71°C | |
Glass Bottle | |
1mbar at 48°C | |
Undesignated | |
500mL | |
112-40-3 | |
C12H26 | |
MFCD00008969 | |
01, 171 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCCCCCCCCCC | |
170.34 | |
CHEBI:28817 | |
Liquid | |
Pure |
0.7530g/mL | |
Authentic | |
1.4210 to 1.4230 | |
215.0°C to 217.0°C | |
-12.0°C | |
99% | |
99% | |
CH3(CH2)10CH3 | |
2810 | |
adakane 12, alkanes, c10-14, bihexyl, ccris 661, dihexyl, duodecane, n-dodecan, n-dodecan german, n-dodecane, undecane, methyl | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
dodecane | |
8182 | |
170.34 | |
99% |
Chemical Identifiers
112-40-3 | |
170.34 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
8182 | |
dodecane |
C12H26 | |
MFCD00008969 | |
adakane 12, alkanes, c10-14, bihexyl, ccris 661, dihexyl, duodecane, n-dodecan, n-dodecan german, n-dodecane, undecane, methyl | |
CHEBI:28817 | |
CCCCCCCCCCCC |
Safety and Handling
GHS P Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Combustible liquid. Harmful by inhalation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking.
NFPA
Danger
EINECSNumber : 203-967-9
RTECSNumber : JR2125000
TSCA : TSCA