missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Dodecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 434590010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Dodecane | |
-12.0°C | |
0.7530g/mL | |
71°C | |
98.5% min. (GC) | |
C12H26 | |
CH3(CH2)10CH3 | |
0.753 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCCCCCCCCCC | |
170.34 | |
<7 mm2/s (40°C) | |
170.34 | |
Liquid |
112-40-3 | |
Colorless | |
215.0°C to 217.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.4210 to 1.4230 | |
1 L | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
dodecane | |
8182 | |
CHEBI:28817 | |
99% |
Chemical Identifiers
112-40-3 | |
170.34 | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
CHEBI:28817 | |
CCCCCCCCCCCC |
C12H26 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
8182 | |
dodecane |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 203-967-9
RUO – Research Use Only