Learn More
Alfa Aesar™ Dodecacarbonyltriosmium, 99%
Manufacturer: Alfa Aesar™ 1303802
Specifications
Dodecacarbonyltriosmium | |
0.5g | |
C12O12Os3 | |
UN3466 | |
VUBLMKVEIPBYME-UHFFFAOYSA-N | |
carbon monoxide;osmium | |
6096995 | |
Crystalline |
224°C | |
15696-40-9 | |
MFCD00011149 | |
osmium carbonyl, triangulo-dodecacarbonyltriosmium, triosmium dodecacarbonyl, tri-osmium dodecacarbonyl, triosmium dodecacarbonyl | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Os].[Os].[Os] | |
906.81 | |
906.73 | |
99% |
Chemical Identifiers
15696-40-9 | |
906.81 | |
VUBLMKVEIPBYME-UHFFFAOYSA-N | |
6096995 | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Os].[Os].[Os] |
C12O12Os3 | |
MFCD00011149 | |
osmium carbonyl, triangulo-dodecacarbonyltriosmium, triosmium dodecacarbonyl, tri-osmium dodecacarbonyl, triosmium dodecacarbonyl | |
carbon monoxide;osmium |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.