missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dodecacarbonyltetracobalt, 98% min

Manufacturer:  Alfa Aesar™ 1305506

 View more versions of this product

Catalog No. AA1305506



Specifications

Specifications

Dodecacarbonyltetracobalt
C12Co4O12
MFCD00064698
dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]
571.853
571.86
≤98%
Air Sensitive
60°C (dec.)
17786-31-1
Co4(CO)12
UN3179
YMFAWOSEDSLYSZ-UHFFFAOYSA-N
carbon monoxide;cobalt
6096999
Crystalline
2.09g/mL
Black
5g

Chemical Identifiers

17786-31-1
571.853
YMFAWOSEDSLYSZ-UHFFFAOYSA-N
6096999
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]
C12Co4O12
MFCD00064698
dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min
carbon monoxide;cobalt
Safety and Handling

Safety and Handling

GHS H Statement
H228-H301-H331-H351-H317
Flammable solid.
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P280h-P262-P304+P340-P309-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.

RTECSNumber : GG3410000

Documents