missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Doctors Solution, Sodium Plumbite, Certified, 3.2 to 3.5N, LabChem™

Manufacturer:  LabChem LC136902

 View more versions of this product

Catalog No. LC136902



Specifications

Specifications

Doctors Solution
86.4,11.3,2.3
3.1 to 3.3N at pH 11.0
3.2 to 3.5N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Liquid
Passes Test
Soluble in water
Room Temperature
1.8cSt
Colorless
1L
Sodium Plumbite
1310-73-2,1317-36-8
PbO
1.1g/mL
223.19
Certified
Poly Bottle
1.1g/mL
Traceable to NIST
Fume; Lead oxide; Thermal decomposition generates: Corrosive vapors
7.0
Documents
Promotions

Promotions