Please login to your online account to display your discounted pricing

Docosanoic acid, 85%, Tech., ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC334812500

 View more versions of this product

Catalog No. AC334812500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥80%
Physical Form Crystalline Powder
Formula Weight 340.58
Purity Grade Technical Grade
Quantity 250g
Color White to slightly yellow
Beilstein 02, 391
Merck Index 15, 1022
Assay Percent Range 85%
CAS 112-85-6
Chemical Name or Material Docosanoic acid
Molecular Formula C22H44O2
MDL Number MFCD00002807
Packaging Glass bottle
Synonym Behenic acid
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Docosanoic acid
CAS 112-85-6
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C22H44O2
Formula Weight 340.58
MDL Number MFCD00002807
Synonym Behenic acid