missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Docosanal, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H33924MD

 View more versions of this product

Catalog No. AAH33924MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Docosanal
57°C to 60°C
CH3(CH2)20CHO
250 mg
behenoyl, n-docosanal, docosyl aldehyde, 1-docosanal
CCCCCCCCCCCCCCCCCCCCCC=O
324.59
324.59
57402-36-5
C22H44O
MFCD07780595
Air Sensitive
ULCXRAFXRZTNRO-UHFFFAOYSA-N
docosanal
3017068
98%

Chemical Identifiers

57402-36-5
324.59
ULCXRAFXRZTNRO-UHFFFAOYSA-N
3017068
CCCCCCCCCCCCCCCCCCCCCC=O
C22H44O
MFCD07780595
behenoyl, n-docosanal, docosyl aldehyde, 1-docosanal
docosanal
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only