Please login to your online account to display your discounted pricing

Docetaxel, Tocris™

Microtubule stabilizer

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Docetaxel
Assay Percent Range >99%
Molecular Formula C43H53NO14
Formula Weight 807.88
Synonym [2aR-[2aα,4β,4aβ,6β,9α(αR™,βS™),11α,12α,12aβ
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

405610

 
r & d systems
RDS405610
10mg Each for $164.61

405650

 
r & d systems
RDS405650
50mg Each for $682.67
Description & Specifications

Specifications

Formula Weight 807.88
Quantity 10mg
Chemical Name or Material Docetaxel
CAS 114977-28-5
Assay Percent Range >99%
Synonym [2aR-[2aα,4β,4aβ,6β,9α(αR™,βS™),11α,12α,12aβ
Molecular Formula C43H53NO14
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C