missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dobutamine hydrochloride

Manufacturer:  Alfa Aesar™ J6133303

 View more versions of this product

Catalog No. AAJ6133303



Description

Description

Applications
Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist with potent positive inotropic effects in vivo. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1 μM.

Solubility
Soluble in water or ethanol with warming. Also soluble in methanol

Notes
Solutions oxidize readily at pH greater than 11. Protect solutions from light
Specifications

Specifications

Dobutamine hydrochloride
184°C to 186°C
49745-95-1
MFCD00153795
+--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, dobutamine hcl, dobutamine hydrochloride, dobutrex, inotrex
BQKADKWNRWCIJL-UHFFFAOYSA-N
4-[2-[4-(4-hydroxyphenyl)butan-2-ylamino]ethyl]benzene-1,2-diol;hydrochloride
65324
337.85
White
1g
C18H24ClNO3
14,3395
Soluble in water or ethanol with warming; Also soluble in methanol
CC(CCC1=CC=C(C=C1)O)NCCC2=CC(=C(C=C2)O)O.Cl
337.844
CHEBI:4671
Off-white to white powder

Chemical Identifiers

49745-95-1
337.844
BQKADKWNRWCIJL-UHFFFAOYSA-N
65324
4-[2-[4-(4-hydroxyphenyl)butan-2-ylamino]ethyl]benzene-1,2-diol;hydrochloride
C18H24ClNO3
MFCD00153795
+--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, dobutamine hcl, dobutamine hydrochloride, dobutrex, inotrex
CHEBI:4671
CC(CCC1=CC=C(C=C1)O)NCCC2=CC(=C(C=C2)O)O.Cl
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H312-H332
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H361

EINECSNumber : 256-464-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents