Please login to your online account to display your discounted pricing

DO11.10 TCR Mouse anti-Mouse, PE-Cy7, Clone: KJ126, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $462.55

Specifications

Antigen DO11.10 TCR
Clone KJ1-26 (KJ126
Host Species Mouse
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123388

 
life technologies
LS25580880
25μg Each for $187.05

501123389

 
life technologies
LS25580882
100μg Each for $462.55
Description & Specifications

Specifications

Antigen DO11.10 TCR
Clone KJ1-26 (KJ126
Host Species Mouse
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The KJ1-26 anti-idiotypic monoclonal antibody binds the T-cell receptor (TCR) expressed on DO11.10 and DO11.10.24 murine T cell hybridomas, but not on any other T cell hybridomas tested.The TCR expressed on DO11.10 recognizes an ovalbumin peptide in the context of I-Ad major histocompatibility molecules.The use of DO11.10 and KJ1-26 have proven to be instrumental in describing many of the intricate details of T cell immunity.