Please login to your online account to display your discounted pricing

Thermo Scientific™ dNTP Mixes - PROMO

Reduce the number of pipetting steps and the risk of reaction setup errors with these dNTP mixes, premixed aqueous solutions of dATP, dTTP, dCTP, and dGTP

$184.28 - $184.28

Volume 1mL
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

FERR1121PM

 
thermo scientific™
FERR1121
1mL Each for $184.28
Description & Specifications

Specifications

Volume 1mL
Purity >99%
For Use With (Application) For direct use in PCR, reverse transcription and other applications

 dNTP Mixes contain premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2, 10 or 25 mM. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. Mixes offer the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. They are designed for many different molecular biology applications.