Thermo Scientific™ dNTP Mixes - PROMO
Reduce the number of pipetting steps and the risk of reaction setup errors with these dNTP mixes, premixed aqueous solutions of dATP, dTTP, dCTP, and dGTP
Manufacturer:  Thermo Scientific™ FERR1121
 View more versions of this product
Catalog No. FERR1121PM
Specifications
Description
Nucleotides, dNTP;Thermo Scientific; dNTP Mix, 25mm; 1mL
dNTP Mixes contain premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2, 10 or 25 mM. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. Mixes offer the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. They are designed for many different molecular biology applications
Description & Specifications
Specifications
For direct use in PCR, reverse transcription and other applications
1mL
Each mix at a final concentration of 2, 10 or 25mm.
>99%
1mL