Please login to your online account to display your discounted pricing

DNQX, Tocris


Selective non-NMDA antagonist

Quantity 10mg

Manufacturer: r & d systems  018910

 View more versions of this product

Catalog No. 018910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 252.14
CAS 2379-57-9
Chemical Name or Material DNQX
Assay Percent Range >98%
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione
Molecular Formula C8H4N4O6
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material DNQX
Assay Percent Range >98%
Molecular Formula C8H4N4O6
Formula Weight 252.14
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione
Accessories

Store at Room Temperature