Please login to your online account to display your discounted pricing

DNQX, Tocris

Selective non-NMDA antagonist

$73.75 - $233.76

Chemical Identifiers

Chemical Name or Material DNQX
Assay Percent Range >98%
Molecular Formula C8H4N4O6
Formula Weight 252.14
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

018950

 
r & d systems
018950
50mg Each for $233.76

018910

 
r & d systems
018910
10mg Each for $73.75
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 252.14
Chemical Name or Material DNQX
CAS 2379-57-9
Molecular Formula C8H4N4O6
Assay Percent Range >98%
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature