Please login to your online account to display your discounted pricing

DNQX disodium salt, Tocris

Selective non-NMDA antagonist; Water-soluble salt of DNQX

$88.05 - $376.43

Chemical Identifiers

Chemical Name or Material DNQX disodium salt
Assay Percent Range >98%
Molecular Formula C8H2N4O6Na2
Formula Weight 296.1
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

231250

 
r & d systems
231250
50mg Each for $376.43

231210

 
r & d systems
231210
10mg Each for $88.05
Description & Specifications

Specifications

Formula Weight 296.1
Quantity 50mg
Chemical Name or Material DNQX disodium salt
Synonym 6, 7-Dinitroquinoxaline-2, 3-dione disodium salt
CAS 1312992-24-7
Molecular Formula C8H2N4O6Na2
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature