Please login to your online account to display your discounted pricing

Thermo Scientific DNAPac™ PA200 Oligonucleotide Columns


Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide Columns.

Manufacturer: thermo scientific  SP6731

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. DXSP6731

  • / Each
 

Description & Specifications

Specifications

For Use With HPAE-PAD
Particle Size 8µm
Max. Pressure 4000psi
Pore Size Non-porous
pH 2.5 to 12.5
Surface Area -
Temperature 85°C
Carbon Load -
Stationary Phase Quaternary ammonium
Packing Material Spherical, non-porous polymer
Type Guard Column
Column Format Guard Column
Diameter (Metric) 22mm
USP Type N/A
Column Type Ion Exchange
Length (Metric) 50mm
Endcapped -

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide Columns.

  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA