missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA Polymerase, β Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reprucible results in western blotting experiments with Thermo Scientific™ DNA Polymerase, β Ab-2, Mouse Monoclonal Antibody.

$305.45 - $1,501.35

Specifications

Antigen DNA Polymerase, beta Ab-2
Clone 61
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1402P Thermo Scientific™ Lab Vision™
MS1402P
200μg/mL 1mL Each for $1,404.86
MS1402P0 Thermo Scientific™ Lab Vision™
MS1402P0
200μg/mL 100μL Each for $305.45
MS1402P1 Thermo Scientific™ Lab Vision™
MS1402P1
200μg/mL 500μL Each for $839.61
MS1402P1ABX Thermo Scientific™ Lab Vision™
MS1402P1ABX
1mg/mL 100μL Each for $959.45
MS1402PABX Thermo Scientific™ Lab Vision™
MS1402PABX
1mg/mL 200μL Each for $1,501.35
Specifications

Description

Obtain accurate, reprucible results in western blotting experiments with Thermo Scientific™ DNA Polymerase, β Ab-2, Mouse Monoclonal Antibody.

DNA polymerase Β comprises an amino-terminal 8-kDa domain and a carboxy-terminal 31-kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase Β, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates β-pol level. DNA polymerase Β fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.

Host Species: Mouse

Clone: 61

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: DNA polymerase β protein

Molecular Weight: 39kDa

Positive Control: HeLa cells, Testis

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

DNA Polymerase, beta Ab-2
Western Blot
Unlabeled
Murine
DNA Repair
DNA polymerase beta protein
61
Monoclonal
Concentrated
RUO
Primary
IgG2a