Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA Polymerase, β Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, β Ab-1, Mouse Monoclonal Antibody.

$288.92 - $1,686.35

Specifications

Antigen DNA Polymerase, beta Ab-1
Clone 18S
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS669B

 
thermo scientific™ lab vision™
MS669B
200μg/mL Biotin 1mL Each for $1,686.35

MS669B0

 
thermo scientific™ lab vision™
MS669B0
200μg/mL Biotin 100μL Each for $386.65

MS669B1

 
thermo scientific™ lab vision™
MS669B1
200μg/mL Biotin 500μL Each for $1,053.00

MS669P

 
thermo scientific
MS669P
200μg/mL Unlabeled 1mL Each for $1,256.04

MS669P0

 
thermo scientific™ lab vision™
MS669P0
200μg/mL Unlabeled 100μL Each for $288.92

MS669P1

 
thermo scientific™ lab vision™
MS669P1
200μg/mL Unlabeled 500μL Each for $810.80

MS669P1ABX

 
thermo scientific™ lab vision™
MS669P1ABX
1mg/mL Unlabeled 100μL Each for $883.67

MS669PABX

 
thermo scientific™ lab vision™
MS669PABX
1mg/mL Unlabeled 200μL Each for $1,470.02
Description & Specifications

Specifications

Antigen DNA Polymerase, beta Ab-1
Clone 18S
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Rat DNA polymerase beta protein
Isotype IgG1

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, β Ab-1, Mouse Monoclonal Antibody.

DNA polymerase Β comprises an amino-terminal 8-kDa domain and a carboxy-terminal 31-kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase Β, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates β-pol level. DNA polymerase Β fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.

Host Species: Mouse

Clone: 18S

Isotype: IgG1

Species Reactivity: Human, Cow, Mouse, Rat, Hamster and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Rat DNA polymerase β protein

Molecular Weight: 39 kDa

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Effect on Polymerase Activity (Noninhibitory)

Note:

USA: RUO; Int'l: RUO