Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS576P1ABX

 View more versions of this product

Catalog No. MS576P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Format Concentrated
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Antigen DNA Polymerase, epsilon Ab-1
Immunogen Purified recombinant human DNA polymerase epsilon corresponding to aa1-176
Isotype IgG2a kappa
Host Species Mouse
Clone 93H3A
Conjugate Unlabeled
Research Discipline DNA Repair
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody.

Polε is present in proliferating cells and is implicated in repair of UV-damaged DNA.

Host Species: Mouse

Clone: 93H3A

Isotype: IgG1/κ

Species Reactivity: Detects both polε and polε of Human, Monkey, Cow and Mouse. A mixture of Ab-1 & Ab-2 reacts with D. melanogaster (polε only), T. ni insect cells (polε only), S. cerevisiae (both polε and polε), X. laevis (polε)

Epitope: aa 1-176

Immunogen: Purified recombinant human DNA polymerase ε corresponding to aa1-176

Molecular Weight: >200kDa identified as DNA polymerase ε (polε) and 140 kDa polε

Positive Control: 293 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO