Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody.

$276.63 - $1,505.10

Specifications

Antigen DNA Polymerase, epsilon Ab-1
Clone 93H3A
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS576B

 
thermo scientific™ lab vision™
MS576B
200μg/mL Biotin 1mL Each for $1,505.10

MS576B0

 
thermo scientific™ lab vision™
MS576B0
200μg/mL Biotin 100µL Each for $375.18

MS576B1

 
thermo scientific™ lab vision™
MS576B1
200μg/mL Biotin 500µL Each for $1,020.86

MS576P

 
thermo scientific™ lab vision™
MS576P
200μg/mL Unlabeled 1mL Each for $1,179.18

MS576P0

 
thermo scientific™ lab vision™
MS576P0
200μg/mL Unlabeled 100µL Each for $276.63

MS576P1

 
thermo scientific™ lab vision™
MS576P1
200μg/mL Unlabeled 500µL Each for $785.18

MS576P1ABX

 
thermo scientific™ lab vision™
MS576P1ABX
1mg/mL Unlabeled 100µL Each for $871.94

MS576PABX

 
thermo scientific™ lab vision™
MS576PABX
1mg/mL Unlabeled 200µL Each for $1,240.76
Description & Specifications

Specifications

Antigen DNA Polymerase, epsilon Ab-1
Clone 93H3A
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Purified recombinant human DNA polymerase epsilon corresponding to aa1-176
Isotype IgG2a kappa

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ DNA Polymerase, ε Ab-1, Mouse Monoclonal Antibody.

Polε is present in proliferating cells and is implicated in repair of UV-damaged DNA.

Host Species: Mouse

Clone: 93H3A

Isotype: IgG1/κ

Species Reactivity: Detects both polε and polε of Human, Monkey, Cow and Mouse. A mixture of Ab-1 & Ab-2 reacts with D. melanogaster (polε only), T. ni insect cells (polε only), S. cerevisiae (both polε and polε), X. laevis (polε)

Epitope: aa 1-176

Immunogen: Purified recombinant human DNA polymerase ε corresponding to aa1-176

Molecular Weight: >200kDa identified as DNA polymerase ε (polε) and 140 kDa polε

Positive Control: 293 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO