Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-4 (Cocktail), Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.

$272.78 - $1,403.14

Specifications

Antigen DNA-PKcs Ab-4 (Cocktail)
Clone 18-2 + 25-4 + 42-psc
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS423P

 
thermo scientific™ lab vision™
MS423P
200μg/mL 1mL Each for $1,283.47

MS423P0

 
thermo scientific™ lab vision™
MS423P0
200μg/mL 100µL Each for $272.78

MS423P1

 
thermo scientific™ lab vision™
MS423P1
200μg/mL 500µL Each for $766.43

MS423P1ABX

 
thermo scientific™ lab vision™
MS423P1ABX
1mg/mL 100µL Each for $835.10

MS423PABX

 
thermo scientific™ lab vision™
MS423PABX
1mg/mL 200µL Each for $1,403.14
Description & Specifications

Specifications

Antigen DNA-PKcs Ab-4 (Cocktail)
Clone 18-2 + 25-4 + 42-psc
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Isotype IgG1 IgG2a

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments with Thermo Scientific™ DNA-PKcs Ab-4 (Cocktail), Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2 + 25-4 + 42-psc

Isotype: IgG1 + IgG2a + IgG1

Species Reactivity: Human, Mouse, Hamster, Xenopus and Chicken. Others not known.

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil or testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO