Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-4 (Cocktail), Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.

Manufacturer: thermo scientific™ lab vision™  MS423P1ABX

 View more versions of this product

Catalog No. MS423P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1 IgG2a
Antigen DNA-PKcs Ab-4 (Cocktail)
Format Concentrated
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Host Species Mouse
Regulatory Status RUO
Clone 18-2 + 25-4 + 42-psc
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline DNA Repair

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments with Thermo Scientific™ DNA-PKcs Ab-4 (Cocktail), Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2 + 25-4 + 42-psc

Isotype: IgG1 + IgG2a + IgG1

Species Reactivity: Human, Mouse, Hamster, Xenopus and Chicken. Others not known.

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil or testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO