Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and gel supershift experiments.

$522.76 - $1,755.73

Specifications

Antigen DNA-PKcs Ab-3
Clone 42-psc; same as 42-26 or 42-27
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS371P

 
thermo scientific™ lab vision™
MS371P
200μg/mL 1mL Each for $1,597.27

MS371P0

 
thermo scientific™ lab vision™
MS371P0
200μg/mL 100µL Each for $522.76

MS371P1

 
thermo scientific™ lab vision™
MS371P1
200μg/mL 500µL Each for $1,102.73

MS371P1ABX

 
thermo scientific™ lab vision™
MS371P1ABX
1mg/mL 100µL Each for $1,239.88

MS371PABX

 
thermo scientific™ lab vision™
MS371PABX
1mg/mL 200µL Each for $1,755.73
Description & Specifications

Specifications

Antigen DNA-PKcs Ab-3
Clone 42-psc; same as 42-26 or 42-27
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Isotype IgG2a kappa

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and gel supershift experiments with Thermo Scientific™ DNA-PKcs Ab-3, Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 42-psc; same as 42-26 or 42-27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 2713-3198

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: Testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO